Anti-COL6A2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-403
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:COL6A2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P12110
- Antigen Synonyms:CO6A2_HUMAN|Collagen alpha 2(VI) chain|Collagen alpha-2(VI) chain|collagen type VI alpha 2|Collagen VI alpha 2 polypeptide|human mRNA for collagen VI alpha 2 C terminal globular domain|PP3610
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Collagen alpha-2(VI) chain protein (254-400AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: COL6A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human