Anti-DAP3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:10 to 1:50
Type:
Antigen: DAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-550
- 77847-549
- Antigen Symbol:
- DAP3
- DAP3
- Antigen Synonyms:
- 28S ribosomal protein S29|28S ribosomal protein S29 mitochondrial|bMRP10|DAP 3|DAP-3|Dap3|Death associated protein 3|Death-associated protein 3|DKFZp686G12159|FLJ12817|Ionizing radiation resistance conferring protein|MGC126058|MGC126059|Mitochondrial 28S ribosomal protein S29|mitochondrial|MITOCHONDRIAL RIBOSOMAL PROTEIN S29|MRP S29|MRP-S29|MRPS29|OTTHUMP00000033480|OTTHUMP00000033481|PSD-95/SAP90-binding protein 3|RT29_HUMAN|S29mt|SAP90/PSD-95-associated protein 3|SAPAP3
- 28S ribosomal protein S29|28S ribosomal protein S29 mitochondrial|bMRP10|DAP 3|DAP-3|Dap3|Death associated protein 3|Death-associated protein 3|DKFZp686G12159|FLJ12817|Ionizing radiation resistance conferring protein|MGC126058|MGC126059|Mitochondrial 28S ribosomal protein S29|mitochondrial|MITOCHONDRIAL RIBOSOMAL PROTEIN S29|MRP S29|MRP-S29|MRPS29|OTTHUMP00000033480|OTTHUMP00000033481|PSD-95/SAP90-binding protein 3|RT29_HUMAN|S29mt|SAP90/PSD-95-associated protein 3|SAPAP3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DAP3
- Fusion protein of Human DAP3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together