Anti-ZNRF3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNRF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-096
- 77844-095
- Antigen Symbol:
- ZNRF3
- ZNRF3
- Antigen Synonyms:
- KIAA1133|RNF203|E3 ubiquitin-protein ligase EC 2.3.2.27|RING finger protein 203|RING-type E3 ubiquitin transferase Zinc/RING finger protein 3
- KIAA1133|RNF203|E3 ubiquitin-protein ligase EC 2.3.2.27|RING finger protein 203|RING-type E3 ubiquitin transferase Zinc/RING finger protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULT6
- Q9ULT6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human E3 ubiquitin-protein ligase ZNRF3 protein (56-219AA)
- Recombinant human E3 ubiquitin-protein ligase ZNRF3 protein (56-219AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together