Anti-MCHR1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000, IHC:1:50 to 1:200
Type:
Antigen: MCHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77850-092
- 77850-091
- Antigen Symbol:
- MCHR1
- MCHR1
- Antigen Synonyms:
- MCHR1|GPR24|SLC1Melanin-concentrating hormone receptor 1|MCH receptor 1|MCH-R1|MCHR-1|G-protein coupled receptor 24|MCH-1R|MCH1R|MCHR|SLC-1|Somatostatin receptor-like protein
- MCHR1|GPR24|SLC1Melanin-concentrating hormone receptor 1|MCH receptor 1|MCH-R1|MCHR-1|G-protein coupled receptor 24|MCH-1R|MCH1R|MCHR|SLC-1|Somatostatin receptor-like protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99705
- Q99705
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MCHR1
- Synthetic peptide of Human MCHR1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together