Anti-HNRNPM Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77849-839
- 77849-840
- Antigen Symbol:
- HNRNPM
- HNRNPM
- Antigen Synonyms:
- CEA receptor|CEAR|Heterogeneous nuclear ribonucleoprotein M|Heterogenous nuclear ribonucleoprotein M4|hnRNA binding protein M4|hnRNP M|Hnrnpm|HNRNPM4|HNRPM|HNRPM_HUMAN|HNRPM4|HTGR1|M3|M4|M4 protein|N-acetylglucosamine receptor 1|NAGR1
- CEA receptor|CEAR|Heterogeneous nuclear ribonucleoprotein M|Heterogenous nuclear ribonucleoprotein M4|hnRNA binding protein M4|hnRNP M|Hnrnpm|HNRNPM4|HNRPM|HNRPM_HUMAN|HNRPM4|HTGR1|M3|M4|M4 protein|N-acetylglucosamine receptor 1|NAGR1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52272
- P52272
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human HNRNPM
- Synthetic peptide of Human HNRNPM
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together