Anti-RAB26 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:100 to 1:300
Type:
Antigen: RAB26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-812
- 77849-811
- Antigen Symbol:
- RAB26
- RAB26
- Antigen Synonyms:
- CATX-8|RAB11c|RAB26 member RAS oncogene family|RAB26_HUMAN|Ras related oncogene protein|Ras related protein Rab 26|Ras-associated protein Ras-related protein Rab-26|V46133
- CATX-8|RAB11c|RAB26 member RAS oncogene family|RAB26_HUMAN|Ras related oncogene protein|Ras related protein Rab 26|Ras-associated protein Ras-related protein Rab-26|V46133
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULW5
- Q9ULW5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human RAB26
- Synthetic peptide of Human RAB26
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together