Anti-CD3EAP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77852-665
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD3EAP
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O15446
- Antigen Synonyms:A34.5|Anti sense to ERCC 1 protein|Antisense to ERCC-1 protein|ASE 1|ASE-1|ASE1|CAST|CD3 epsilon associated protein|CD3-epsilon-associated protein|CD3E antigen|epsilon polypeptide associated protein|CD3E associated protein|CD3e molecule|epsilon associated protein|CD3E-associated protein|CD3EAP|DNA directed RNA polymerase I subunit RPA34|DNA-directed RNA polymerase I subunit RPA34|MGC118851|PAF 49|PAF49|RNA polymerase I associated factor PAF49|RNA polymerase I-associated factor PAF49|RPA34_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA-directed RNA polymerase I subunit RPA34 protein (289-404AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CD3EAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human