Anti-REG4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: REG4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-002
- 77843-003
- Antigen Symbol:
- REG4
- REG4
- Antigen Synonyms:
- Gastrointestinal secretory protein|GISP|Reg IV|REG like protein|REG-4|REG-like protein|REG4_HUMAN|Regenerating gene type IV|Regenerating islet derived family member 4|Regenerating islet derived protein 4 precursor|Regenerating islet-derived protein 4|Regenerating islet-derived protein IV|REGIV|RELP
- Gastrointestinal secretory protein|GISP|Reg IV|REG like protein|REG-4|REG-like protein|REG4_HUMAN|Regenerating gene type IV|Regenerating islet derived family member 4|Regenerating islet derived protein 4 precursor|Regenerating islet-derived protein 4|Regenerating islet-derived protein IV|REGIV|RELP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYZ8
- Q9BYZ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Regenerating islet-derived protein 4 protein (27-148AA)
- Recombinant Human Regenerating islet-derived protein 4 protein (27-148AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together