Anti-EPHA6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: EPHA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-797
- 77843-796
- Antigen Symbol:
- EPHA6
- EPHA6
- Antigen Synonyms:
- Ehk 2|EHK-2|Ehk2|EPH homology kinase 2|Eph Receptor A6|EPHA6_HUMAN|ephrin receptor ephrin type A receptor 6|Ephrin type-A receptor 6|Hek12
- Ehk 2|EHK-2|Ehk2|EPH homology kinase 2|Eph Receptor A6|EPHA6_HUMAN|ephrin receptor ephrin type A receptor 6|Ephrin type-A receptor 6|Hek12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UF33
- Q9UF33
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Ephrin type-A receptor 6 protein (132-279AA)
- Recombinant human Ephrin type-A receptor 6 protein (132-279AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together