Anti-HK1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-447
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HK1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P19367
- Antigen Synonyms:BB404130|Brain form hexokinase|dea|DrHXK1|EC 2.7.1.1|Glycolytic enzyme|HEXOKIN|hexokinase I|Hexokinase PI|Hexokinase type I|Hexokinase|tumor isozyme|Hexokinase-1|Hexokinase-A|HK I|HK1 tb|HK1 tc|Hk1-s|HK1-ta|HK1-tb|HK1-tc|HKD|HKI|HMSNR|HXK1|HXK1_HUMAN|im:7148527|mHk1-s|wu:fc09d08|wu:fc16e02|wu:fc21e02|wu:fq14b11|zgc:55790|zgc:77618
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Hexokinase-1 protein (13-475AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: HK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat