Anti-CRHR1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-550
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRHR1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:P34998
- Antigen Synonyms:Corticotropin releasing factor type 1 receptor|corticotropin releasing hormone receptor 1|Corticotropin-releasing factor receptor 1|Corticotropin-releasing hormone receptor 1|CRF R|crf receptor 1|crf receptor type 1|crf type 1|CRF-R|CRF-R-1|CRF-R1|CRF1|CRFR|CRFR-1|CRFR1|CRFR1_HUMAN|CRH-R-1|CRH-R1|CRH-R1h|CRHR|CRHR1f|CRHR1L
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Corticotropin-releasing factor receptor 1 protein (24-121AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CRHR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human