Anti-Adrb2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-832
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Adrb2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P10608
- Antigen Synonyms:Adrb2r|Beta-2 adrenergic receptor|Beta-2 adrenoreceptor|Beta-2 adrenoceptor
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Rat Beta-2 adrenergic receptor protein (330-418AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: Adrb2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat