Order Entry
United States
ContactUsLinkComponent
Anti-NIP7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-NIP7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-NIP7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77846-052
  • 77846-053
  • Antigen Symbol:
  • NIP7
  • NIP7
  • Antigen Synonyms:
  • 60S ribosome subunit biogenesis protein NIP7 homolog|CGI 37|CGI37|FLJ10296|HSPC031|HSPC180|KD93|NIP 7|NIP7|NIP7 nucleolar pre rRNA processing protein|NIP7_HUMAN|Nuclear import 7|Nuclear import 7 homolog (S. cerevisiae)|Nuclear import 7 homolog|OK/SW cl.76|OK/SW cl.78|OTTHUMP00000174889
  • 60S ribosome subunit biogenesis protein NIP7 homolog|CGI 37|CGI37|FLJ10296|HSPC031|HSPC180|KD93|NIP 7|NIP7|NIP7 nucleolar pre rRNA processing protein|NIP7_HUMAN|Nuclear import 7|Nuclear import 7 homolog (S. cerevisiae)|Nuclear import 7 homolog|OK/SW cl.76|OK/SW cl.78|OTTHUMP00000174889
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y221
  • Q9Y221
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human 60S ribosome subunit biogenesis protein NIP7 homolog protein (1-180AA)
  • Recombinant Human 60S ribosome subunit biogenesis protein NIP7 homolog protein (1-180AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95% Protein G purified
  • >95% Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together