Anti-ADH1C Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-779
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADH1C
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P00326
- Antigen Synonyms:ADH gamma subunit|ADH1G_HUMAN|Adh3|Alcohol dehydrogenase 1C (class I) gamma polypeptide|Alcohol dehydrogenase 1C|Alcohol dehydrogenase 1C gamma polypeptide|Alcohol dehydrogenase 3 (class I) gamma polypeptide|Alcohol dehydrogenase 3|Alcohol dehydrogenase subunit gamma|Aldehyde reductase|Class I alcohol dehydrogenase|OTTHUMP00000220209
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alcohol dehydrogenase 1C protein (1-375AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ADH1C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human