Anti-AMPD1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77851-111
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AMPD1
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P23109
- Antigen Synonyms:AMPD1AMP deaminase 1|EC 3.5.4.6|AMP deaminase isoform M|Myoadenylate deaminase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human AMP deaminase 1 protein (97-272AA)
- Purification:>95%, Protein G purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human