Anti-BTAF1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-961
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BTAF1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O14981
- Antigen Synonyms:ATP dependent helicase ATP-dependent helicase B-TFIID transcription factor associated 170 kDa subunit|B-TFIID transcription factor associated, 170kDa (Mot1 homolog, S. cerevisiae)|B-TFIID transcription factor-associated 170 kDa subunit|BTAF 1 RNA polymerase II|BTAF1 RNA polymerase II B TFIID transcription factor associated 170kDa|BTAF1_HUMAN|MOT 1|MOT1|MOT1 Yeast homolog of|TAF 172|TAF II 170|TAF(II)170|TAF-172|TAF172|TAFII170|TATA binding protein associated factor 172|TATA-binding protein-associated factor 172|TBP associated factor 172|TBP-associated factor 172
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human TATA-binding protein-associated factor 172 protein (1750-1849AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: BTAF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human