Anti-TPM2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: TPM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77848-135
- 77848-134
- Antigen Symbol:
- TPM2
- TPM2
- Antigen Synonyms:
- Alpha tropomyosin|AMCD1|Arthrogryposis multiplex congenital distal type 1|Beta tropomyosin|Beta-tropomyosin|Cytoskeletal tropomyosin TM30|DA1|DA2B|epididymis secretory protein Li 273|FLJ41118|Heat stable cytoskeletal protein 30 kDa|HEL-S-273|hscp30|HTM alpha|hTM5|MGC14582|MGC3261|MGC72094|NEM1|NEM4|Nemaline myopathy type 4|OK/SW cl.5|Sarcomeric tropomyosin kappa|TM 5|TM3|TM30|TM30nm|TMSA|TMSB|TPM 1|TPM 3|TPM1 alpha|TPM1 kappa|TPM2|TPM2_HUMAN|TRK|Tropomyosin 1 alpha|Tropomyosin 1 alpha chain|Tropomyosin 1 alpha chain isoform 6|Tropomyosin 2 (beta)|Tropomyosin 2|Tropomyosin 3|Tropomyosin alpha 3 chain|Tropomyosin alpha striated muscle isoform|Tropomyosin beta chain|Tropomyosin gamma|Tropomyosin skeletal muscle beta|Tropomyosin-2
- Alpha tropomyosin|AMCD1|Arthrogryposis multiplex congenital distal type 1|Beta tropomyosin|Beta-tropomyosin|Cytoskeletal tropomyosin TM30|DA1|DA2B|epididymis secretory protein Li 273|FLJ41118|Heat stable cytoskeletal protein 30 kDa|HEL-S-273|hscp30|HTM alpha|hTM5|MGC14582|MGC3261|MGC72094|NEM1|NEM4|Nemaline myopathy type 4|OK/SW cl.5|Sarcomeric tropomyosin kappa|TM 5|TM3|TM30|TM30nm|TMSA|TMSB|TPM 1|TPM 3|TPM1 alpha|TPM1 kappa|TPM2|TPM2_HUMAN|TRK|Tropomyosin 1 alpha|Tropomyosin 1 alpha chain|Tropomyosin 1 alpha chain isoform 6|Tropomyosin 2 (beta)|Tropomyosin 2|Tropomyosin 3|Tropomyosin alpha 3 chain|Tropomyosin alpha striated muscle isoform|Tropomyosin beta chain|Tropomyosin gamma|Tropomyosin skeletal muscle beta|Tropomyosin-2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07951
- P07951
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TPM2
- Synthetic peptide of Human TPM2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together