Anti-USP24 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-990
- 77843-991
- Antigen Symbol:
- USP24
- USP24
- Antigen Synonyms:
- Deubiquitinating enzyme 24|KIAA1057|Ubiquitin carboxyl terminal hydrolase 24|Ubiquitin carboxyl-terminal hydrolase 24|Ubiquitin specific peptidase 24|Ubiquitin specific processing protease 24|Ubiquitin specific protease 24|Ubiquitin thioesterase 24|Ubiquitin thiolesterase 24|Ubiquitin-specific-processing protease 24|UBP24_HUMAN|Usp24
- Deubiquitinating enzyme 24|KIAA1057|Ubiquitin carboxyl terminal hydrolase 24|Ubiquitin carboxyl-terminal hydrolase 24|Ubiquitin specific peptidase 24|Ubiquitin specific processing protease 24|Ubiquitin specific protease 24|Ubiquitin thioesterase 24|Ubiquitin thiolesterase 24|Ubiquitin-specific-processing protease 24|UBP24_HUMAN|Usp24
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPU5
- Q9UPU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin carboxyl-terminal hydrolase 24 protein (2509-2620AA)
- Recombinant Human Ubiquitin carboxyl-terminal hydrolase 24 protein (2509-2620AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together