Anti-DQX1 Rabbit Polyclonal Antibody
Catalog # 77846-982
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DQX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q8TE96
- Antigen Synonyms:ATP dependent RNA helicase ATP-dependent RNA helicase DEAQ box 1|DEAQ box polypeptide 1 (RNA dependent ATPase)|DEAQ box polypeptide 1|DEAQ RNA dependent ATPase|DQX1_HUMAN|EC3.6.1|FLJ23757
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human DQX1.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: DQX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse