Anti-Fgf23 Rabbit Recombinant polyclonal Antibody
Catalog # 77855-357
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Fgf23
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8VI82
- Antigen Synonyms:Fgf23Fibroblast growth factor 23|FGF-23
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Rat Fibroblast growth factor 23 protein (25-251AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: Fgf23
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat