Anti-ZCCHC4 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZCCHC4
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-163
- 77843-164
- Antigen Symbol:
- ZCCHC4
- ZCCHC4
- Antigen Synonyms:
- 4930449I23Rik|DHHC domain containing zinc finger protein|HSPC052|MGC21108|OTTMUSP00000027189|ZCHC4_HUMAN|ZGRF4|Zinc finger CCHC domain containing protein 4|Zinc finger CCHC domain-containing protein 4|Zinc finger CCHC type containing 4|Zinc finger GRF type containing 4|Zinc finger, CCHC domain containing 4
- 4930449I23Rik|DHHC domain containing zinc finger protein|HSPC052|MGC21108|OTTMUSP00000027189|ZCHC4_HUMAN|ZGRF4|Zinc finger CCHC domain containing protein 4|Zinc finger CCHC domain-containing protein 4|Zinc finger CCHC type containing 4|Zinc finger GRF type containing 4|Zinc finger, CCHC domain containing 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H5U6
- Q9H5U6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc finger CCHC domain-containing protein 4 protein (25-229AA)
- Recombinant Human Zinc finger CCHC domain-containing protein 4 protein (25-229AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together