Anti-ATF7 Rabbit Recombinant Polyclonal Antibody
77851-528
New Product
- Antigen Symbol:ATF7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P17544
- Antigen Synonyms:1110012F10Rik|9430065F09Rik|Activating transcription factor 7|AI549878|Atf7 protein|ATF7_HUMAN|ATFA|C130020M04Rik|cAMP-dependent transcription factor ATF-7|Cyclic AMP-dependent transcription factor ATF 7|Cyclic AMP-dependent transcription factor ATF-7|MGC31554|MGC57182|Transcription factor ATF A|Transcription factor ATF-A
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cyclic AMP-dependent transcription factor ATF-7 protein (1-117AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, IHC:1:20 to 1:200
Type:
Antigen: ATF7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human