Anti-C8B Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-175
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:C8B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P07358
- Antigen Synonyms:C8b| CO8B_HUMAN| Complement component 8 beta polypeptide| Complement component 8 subunit beta| Complement component C8 beta chain
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Complement component C8 beta chain protein (55-350AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: C8B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human