Anti-CNN3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: CNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-626
- 77849-625
- Antigen Symbol:
- CNN3
- CNN3
- Antigen Synonyms:
- Acidic calponin 3|acidic isoform|Calponin 3 acidic|Calponin 3|Calponin acidic|Calponin acidic isoform|Calponin|Calponin-3|Calponin3|CNN 3|CNN3_HUMAN|dJ639P13.2.2|OTTHUMP00000012470|OTTHUMP00000012471
- Acidic calponin 3|acidic isoform|Calponin 3 acidic|Calponin 3|Calponin acidic|Calponin acidic isoform|Calponin|Calponin-3|Calponin3|CNN 3|CNN3_HUMAN|dJ639P13.2.2|OTTHUMP00000012470|OTTHUMP00000012471
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15417
- Q15417
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CNN3
- Fusion protein of Human CNN3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together