Anti-POLE4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: POLE4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-419
- 77843-418
- Antigen Symbol:
- POLE4
- POLE4
- Antigen Synonyms:
- DNA polymerase epsilon 4 accessory subunit|DNA polymerase epsilon p12 subunit|DNA polymerase epsilon subunit 4|DNA polymerase epsilon subunit p12|DNA polymerase II subunit 4|DPOE4_HUMAN|p12|Polymerase (DNA directed) epsilon 4 (p12 subunit)|Polymerase (DNA directed) epsilon 4|YHHQ1
- DNA polymerase epsilon 4 accessory subunit|DNA polymerase epsilon p12 subunit|DNA polymerase epsilon subunit 4|DNA polymerase epsilon subunit p12|DNA polymerase II subunit 4|DPOE4_HUMAN|p12|Polymerase (DNA directed) epsilon 4 (p12 subunit)|Polymerase (DNA directed) epsilon 4|YHHQ1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NR33
- Q9NR33
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA polymerase epsilon subunit 4 protein (8-117AA)
- Recombinant Human DNA polymerase epsilon subunit 4 protein (8-117AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together