Anti-MMP17 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: MMP17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-024
- 77848-023
- Antigen Symbol:
- MMP17
- MMP17
- Antigen Synonyms:
- Matrix metalloproteinase 17|Matrix metalloproteinase 17 membrane inserted|Matrix metalloproteinase-17|Membrane type 4 matrix metalloproteinase|Membrane type matrix metalloproteinase 4|Membrane-type matrix metalloproteinase 4|Membrane-type-4 matrix metalloproteinase|MMP 17|MMP-17|Mmp17|MMP17_HUMAN|MT MMP 4|MT MMP4|MT-MMP 4|MT4 MMP|MT4-MMP|MT4MMP|MTMMP 4|MTMMP4
- Matrix metalloproteinase 17|Matrix metalloproteinase 17 membrane inserted|Matrix metalloproteinase-17|Membrane type 4 matrix metalloproteinase|Membrane type matrix metalloproteinase 4|Membrane-type matrix metalloproteinase 4|Membrane-type-4 matrix metalloproteinase|MMP 17|MMP-17|Mmp17|MMP17_HUMAN|MT MMP 4|MT MMP4|MT-MMP 4|MT4 MMP|MT4-MMP|MT4MMP|MTMMP 4|MTMMP4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MMP17
- Synthetic peptide of Human MMP17
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together