Anti-DNAJB5 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77854-255
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DNAJB5
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75953
- Antigen Synonyms:DnaJ (Hsp40) homolog subfamily B member 5|DnaJ homolog subfamily B member 5|DNAJB5|DNJB5_HUMAN|Heat shock cognate 40|Heat shock protein cognate 40|Heat shock protein Hsp40 2|Heat shock protein Hsp40 3|Heat shock protein Hsp40-2|Heat shock protein Hsp40-3|Hsc40|Hsp40 2|KIAA1045
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DnaJ homolog subfamily B member 5 protein (1-344AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DNAJB5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human