Anti-Dcn Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77854-023
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Dcn
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:P28654
- Antigen Synonyms:Decorin|Bone proteoglycan II|PG-S2|PG40
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Mouse Decorin protein (35-354AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: Dcn
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse