Anti-ABI1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: ABI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77848-878
- 77848-879
- Antigen Symbol:
- ABI1
- ABI1
- Antigen Synonyms:
- Abelson interactor 1|Abi-1|ABI1_HUMAN|Abl binding protein 4|Abl interactor 1|Abl interactor protein 1|Abl-binding protein 4|AblBP4|e3B1|Eps8 binding protein|Eps8 SH3 domain binding protein|Eps8 SH3 domain-binding protein|Eps8-binding protein|Hssh3bp1|Interactor protein AblBP4|NAP1|Nap1 binding protein|Nap1-binding protein|Nap1BP|Spectrin SH3 domain binding protein 1|Spectrin SH3 domain-binding protein 1|SSH3BP
- Abelson interactor 1|Abi-1|ABI1_HUMAN|Abl binding protein 4|Abl interactor 1|Abl interactor protein 1|Abl-binding protein 4|AblBP4|e3B1|Eps8 binding protein|Eps8 SH3 domain binding protein|Eps8 SH3 domain-binding protein|Eps8-binding protein|Hssh3bp1|Interactor protein AblBP4|NAP1|Nap1 binding protein|Nap1-binding protein|Nap1BP|Spectrin SH3 domain binding protein 1|Spectrin SH3 domain-binding protein 1|SSH3BP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IZP0
- Q8IZP0
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ABI1
- Synthetic peptide of Human ABI1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together