Anti-HCFC1 Rabbit Polyclonal Antibody
Catalog # 77847-296
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HCFC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:P51610
- Antigen Synonyms:C1 factor| CFF| HCF 1| HCF| HCF C-terminal chain 6| HCF-1| HCF1| HCFC1_HUMAN| HFC1| Host cell factor 1| Host cell factor| Host cell factor C1 (VP16 accessory protein)| Host cell factor C1| MGC70925| MRX3| VCAF| VP16 accessory protein
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Synthesized peptide derived from N-terminal of Human HCFC1.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:100
Type:
Antigen: HCFC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse