Anti-ATP5C1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-610
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP5C1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P36542
- Antigen Synonyms:ATP synthase gamma chain|mitochondrial|ATP synthase subunit gamma|ATP synthase|H+ transporting|mitochondrial F1 complex|gamma polypeptide 1|gamma subunit|ATP5C 1|ATP5C|ATP5CL1|ATPG_HUMAN|F ATPase gamma subunit|F-ATPase gamma subunit|Mitochondrial ATP synthase|gamma subunit 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP synthase subunit gamma, mitochondrial protein (156-266AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ATP5C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human