Anti-CGGBP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CGGBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-803
- 77843-802
- Antigen Symbol:
- CGGBP1
- CGGBP1
- Antigen Synonyms:
- 20 kDa CGG binding protein|20 kDa CGG-binding protein|CGBP1_HUMAN|CGG binding protein 1|CGG triplet repeat binding protein 1|CGG triplet repeat-binding protein 1|CGG-binding protein 1|CGGBP 1|CGGBP|OTTHUMP00000213853|OTTHUMP00000213877|p20 CGG binding protein|p20 CGGBP|p20 CGGBP DNA binding protein|p20-CGGBP DNA-binding protein
- 20 kDa CGG binding protein|20 kDa CGG-binding protein|CGBP1_HUMAN|CGG binding protein 1|CGG triplet repeat binding protein 1|CGG triplet repeat-binding protein 1|CGG-binding protein 1|CGGBP 1|CGGBP|OTTHUMP00000213853|OTTHUMP00000213877|p20 CGG binding protein|p20 CGGBP|p20 CGGBP DNA binding protein|p20-CGGBP DNA-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UFW8
- Q9UFW8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human CGG triplet repeat-binding protein 1 protein (1-167AA)
- Recombinant Human CGG triplet repeat-binding protein 1 protein (1-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together