Anti-APC2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-239
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APC2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O95996
- Antigen Synonyms:Adenomatous polyposis coli protein 2|Adenomatous polyposis coli protein-like|APC-like|apc2|APC2_HUMAN|APCL
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenomatous polyposis coli protein 2 protein (2011-2221AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: APC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human