Anti-ABCD2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ABCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-107
- 77847-106
- Antigen Symbol:
- ABCD2
- ABCD2
- Antigen Synonyms:
- ABC39|Abcd2|ABCD2_HUMAN|Adrenoleukodystrophy-like 1|Adrenoleukodystrophy-related protein|ALDL1|ALDR|ALDRP|ATP-binding cassette sub-family D member 2|hALDR
- ABC39|Abcd2|ABCD2_HUMAN|Adrenoleukodystrophy-like 1|Adrenoleukodystrophy-related protein|ALDL1|ALDR|ALDRP|ATP-binding cassette sub-family D member 2|hALDR
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ABCD2
- Synthetic peptide of Human ABCD2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together