Anti-BCAT2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-803
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BCAT2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O15382
- Antigen Synonyms:BCAM|BCAT 2|BCAT(m)|BCAT2_HUMAN|BCATM|BCT2|Branched chain aminotransferase 2|mitochondrial|Branched-chain-amino-acid aminotransferase|ECA40|Placental protein 18|PP18
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Branched-chain-amino-acid aminotransferase, mitochondrial protein (20-200AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: BCAT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human