Anti-CHSY1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-153
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHSY1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q86X52
- Antigen Synonyms:Carbohydrate synthase 1|Chondroitin glucuronyltransferase 1|Chondroitin glucuronyltransferase II|Chondroitin sulfate synthase 1|Chondroitin synthase 1|CHSS1_HUMAN|CHSY|ChSy-1|CSS1|Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1|N acetylgalactosaminyltransferase II|N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1|N-acetylgalactosaminyltransferase 1|TPBS
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Chondroitin sulfate synthase 1 protein (32-148AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: CHSY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human