Anti-AMD1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77851-033
New Product
- Antigen Symbol:AMD1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P17707
- Antigen Synonyms:Adenosylmethionine decarboxylase 1|AdoMetDC|DCAM_HUMAN|EC 4.1.1.50|FLJ26964|S adenosylmethionine decarboxylase 1|S adenosylmethionine decarboxylase proenzyme|S-adenosylmethionine decarboxylase beta chain|SAMDC
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human S-adenosylmethionine decarboxylase proenzyme protein (167-264AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: AMD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human