Anti-AKAP8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: AKAP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-297
- 77848-296
- Antigen Symbol:
- AKAP8
- AKAP8
- Antigen Synonyms:
- A kinase (PRKA) anchor protein 8|A kinase anchor protein 8|A kinase anchor protein 95|A kinase anchor protein 95 kDa|A kinase anchor protein 95kDa|A-kinase anchor protein 8|A-kinase anchor protein 95 kDa|AKAP 8|AKAP 95|AKAP-8|AKAP-95|Akap8|AKAP8_HUMAN|AKAP95|DKFZp586B1222
- A kinase (PRKA) anchor protein 8|A kinase anchor protein 8|A kinase anchor protein 95|A kinase anchor protein 95 kDa|A kinase anchor protein 95kDa|A-kinase anchor protein 8|A-kinase anchor protein 95 kDa|AKAP 8|AKAP 95|AKAP-8|AKAP-95|Akap8|AKAP8_HUMAN|AKAP95|DKFZp586B1222
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43823
- O43823
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human AKAP8
- Synthetic peptide of Human AKAP8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together