Anti-NEUROG2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500 to 1:5000
Type:
Antigen: NEUROG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77843-097
- 77843-098
- Antigen Symbol:
- NEUROG2
- NEUROG2
- Antigen Synonyms:
- Atoh 4|Atoh4|bHLHa8|Class A basic helix-loop-helix protein 8|Math 4A|Math4A|MGC|MGC46562|NEUROG 2|NEUROG2|Neurogenin-2|Neurogenin2|NGN 2|NGN-2|NGN2|NGN2_HUMAN|Protein atonal homolog 4
- Atoh 4|Atoh4|bHLHa8|Class A basic helix-loop-helix protein 8|Math 4A|Math4A|MGC|MGC46562|NEUROG 2|NEUROG2|Neurogenin-2|Neurogenin2|NGN 2|NGN-2|NGN2|NGN2_HUMAN|Protein atonal homolog 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2A3
- Q9H2A3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Neurogenin-2 protein (194-262AA)
- Recombinant Human Neurogenin-2 protein (194-262AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together