Anti-PDS5B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDS5B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-493
- 77843-492
- Antigen Symbol:
- PDS5B
- PDS5B
- Antigen Synonyms:
- PDS5B|APRIN|AS3|KIAA0979|Sister chromatid cohesion protein PDS5 homolog B|Androgen-induced proliferation inhibitor|Androgen-induced prostate proliferative shutoff-associated protein AS3
- PDS5B|APRIN|AS3|KIAA0979|Sister chromatid cohesion protein PDS5 homolog B|Androgen-induced proliferation inhibitor|Androgen-induced prostate proliferative shutoff-associated protein AS3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NTI5
- Q9NTI5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sister chromatid cohesion protein PDS5 homolog B protein (3-124AA)
- Recombinant Human Sister chromatid cohesion protein PDS5 homolog B protein (3-124AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together