Anti-CD33 Rabbit Recombinant Polyclonal Antibody
77852-648
New Product
- Antigen Symbol:CD33
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P20138
- Antigen Synonyms:CD 33|CD33 antigen (gp67)|CD33 antigen|CD33 molecule|CD33_HUMAN|FLJ00391|gp67|My9|Myeloid cell surface antigen Myeloid cell surface antigen CD33 precursor|Myeloid differentiation antigen p67|Sialic acid binding Ig like lectin 3|Sialic acid binding immunoglobulin like lectin 3|Sialic acid-binding Ig-like lectin 3|SIGLEC 3|Siglec-3|SIGLEC3
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Myeloid cell surface antigen CD33 protein (49-259AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: CD33
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human