Anti-CFHR3 Rabbit Recombinant Polyclonal Antibody
77853-016
New Product
- Antigen Symbol:FGFRL1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q8N441
- Antigen Synonyms:FGF homologous factor receptor|FGF receptor like protein 1|FGF receptor-like protein 1|FGFR 5|FGFR like protein|FGFR-5|FGFR-like protein|FGFR5|Fgfrl1|FGRL1_HUMAN|FHFR|Fibroblast growth factor receptor 5|Fibroblast growth factor receptor like 1|Fibroblast growth factor receptor-like 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Fibroblast growth factor receptor-like 1 protein (403-470AA)
- Purification:>95%, Protein G purified
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human