Anti-TNFAIP8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TNFAIP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77848-097
- 77848-098
- Antigen Symbol:
- TNFAIP8
- TNFAIP8
- Antigen Synonyms:
- GG2 1|Head and neck tumor and metastasis-related protein|MDC-3.13|NDED|NF-kappa-B-inducible DED-containing protein|SCC S2|SCC-S2|SCCS2|TFIP8_HUMAN|TNF alpha-induced protein 8|TNF induced protein|TNF-induced protein GG2-1|TNFAIP 8|Tumor necrosis factor alpha induced protein 8|Tumor necrosis factor alpha-induced protein 8
- GG2 1|Head and neck tumor and metastasis-related protein|MDC-3.13|NDED|NF-kappa-B-inducible DED-containing protein|SCC S2|SCC-S2|SCCS2|TFIP8_HUMAN|TNF alpha-induced protein 8|TNF induced protein|TNF-induced protein GG2-1|TNFAIP 8|Tumor necrosis factor alpha induced protein 8|Tumor necrosis factor alpha-induced protein 8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95379
- O95379
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Full length fusion protein
- Full length fusion protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together