Anti-CRIP2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-561
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRIP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P52943
- Antigen Synonyms:CRIP 2|CRIP|CRIP2_HUMAN|CRP 2|CRP|CRP-2|CRP2|Cysteine rich intestinal protein|Cysteine rich protein 2|Cysteine-rich protein 2|ESP 1|ESP1|ESP1 protein|Hlp|Protein ESP1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Cysteine-rich protein 2 protein (1-208AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CRIP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human