Anti-SYNCRIP Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-427
- 77849-428
- Antigen Symbol:
- SYNCRIP
- SYNCRIP
- Antigen Synonyms:
- cytoplasmic RNA-interacting protein|dJ3J17.2|Glycine and tyrosine rich RNA binding protein|Glycine- and tyrosine-rich RNA-binding protein|GRY RBP|GRY-RBP|GRYRBP|Heterogeneous nuclear ribonucleoprotein Q|hnRNP Q|HNRPQ|HNRPQ_HUMAN|HNRPQ1|NS1 associated protein 1|NS1-associated protein 1|NSAP1|pp68|RP1 3J17.2|Synaptotagmin binding cytoplasmic RNA interacting protein|Synaptotagmin-binding|Syncrip
- cytoplasmic RNA-interacting protein|dJ3J17.2|Glycine and tyrosine rich RNA binding protein|Glycine- and tyrosine-rich RNA-binding protein|GRY RBP|GRY-RBP|GRYRBP|Heterogeneous nuclear ribonucleoprotein Q|hnRNP Q|HNRPQ|HNRPQ_HUMAN|HNRPQ1|NS1 associated protein 1|NS1-associated protein 1|NSAP1|pp68|RP1 3J17.2|Synaptotagmin binding cytoplasmic RNA interacting protein|Synaptotagmin-binding|Syncrip
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60506
- O60506
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SYNCRIP
- Synthetic peptide of Human SYNCRIP
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together