Anti-DNASE2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-284
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DNASE2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O00115
- Antigen Synonyms:Acid DNase|Deoxyribonuclease 2 alpha|Deoxyribonuclease 2|Deoxyribonuclease II alpha|Deoxyribonuclease II|Deoxyribonuclease II lysosomal|Deoxyribonuclease-2-alpha|Deoxyribonuclease2|DeoxyribonucleaseII|DNASE 2|DNASE 2A|DNase II alpha|DNase II lysosomal|DNASE2|DNASE2A|DNaseII|DNL 2|DNL|DNL2|DNS2A_HUMAN|Lysosomal DNase II|R31240 2|R31240_2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Deoxyribonuclease-2-alpha protein (19-152AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DNASE2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human