Anti-USP18 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-318
- 77844-317
- Antigen Symbol:
- USP18
- USP18
- Antigen Synonyms:
- 43 kDa ISG15 specific protease|43 kDa ISG15-specific protease|EC 3.1.2.|hUBP43|Interferon Stimulated Gene 43 kD|ISG15 Specific Processing Protease|ISG15-specific-processing protease|ISG43|Ubiquitin Specific Peptidase 18|Ubiquitin Specific Protease 18 43 kD|Ubiquitin Specific Protease 18|Ubl carboxyl terminal hydrolase 18|Ubl carboxyl-terminal hydrolase 18|Ubl thioesterase 18|Ubl thiolesterase 18|Ubp15|UBP18_HUMAN|USP18
- 43 kDa ISG15 specific protease|43 kDa ISG15-specific protease|EC 3.1.2.|hUBP43|Interferon Stimulated Gene 43 kD|ISG15 Specific Processing Protease|ISG15-specific-processing protease|ISG43|Ubiquitin Specific Peptidase 18|Ubiquitin Specific Protease 18 43 kD|Ubiquitin Specific Protease 18|Ubl carboxyl terminal hydrolase 18|Ubl carboxyl-terminal hydrolase 18|Ubl thioesterase 18|Ubl thiolesterase 18|Ubp15|UBP18_HUMAN|USP18
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UMW8
- Q9UMW8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubl carboxyl-terminal hydrolase 18 protein (18-118AA)
- Recombinant Human Ubl carboxyl-terminal hydrolase 18 protein (18-118AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together