Anti-B4GAT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-743
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:B4GAT1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O43505
- Antigen Synonyms:3-N-acetylglucosaminyltransferase 1|3-N-acetylglucosaminyltransferase|B3GN-T1|B3GN1_HUMAN|B3GNT1|B3GNT6|beta-1,3-N-acetylglucosaminyltransferase bGnT-6|BETA3GNTI|I-beta-1|i-beta-1,3-N-acetylglucosaminyltransferase|iGAT|iGnT|N-acetyllactosaminide beta-1|N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase|OTTHUMP00000235058|Poly-N-acetyllactosamine extension enzyme|UDP-GlcNAc:betaGal beta-1|UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1|UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-1,4-glucuronyltransferase 1 protein (161-415AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: B4GAT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human