Anti-MEMO1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MEMO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-400
- 77845-399
- Antigen Symbol:
- MEMO1
- MEMO1
- Antigen Synonyms:
- C21orf19-like protein|C2orf4|CGI 27|DKFZp434I0135|FLJ25031|HCV NS5A-transactivated protein 7|Hepatitis C virus NS5A-transactivated protein 7|Mediator of cell motility 1|Mediator of ErbB2-driven cell motility 1|MEMO|Memo-1|memo1|MEMO1_HUMAN|NS5ATP7|Protein MEMO1
- C21orf19-like protein|C2orf4|CGI 27|DKFZp434I0135|FLJ25031|HCV NS5A-transactivated protein 7|Hepatitis C virus NS5A-transactivated protein 7|Mediator of cell motility 1|Mediator of ErbB2-driven cell motility 1|MEMO|Memo-1|memo1|MEMO1_HUMAN|NS5ATP7|Protein MEMO1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y316
- Q9Y316
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein MEMO1 protein (1-297AA)
- Recombinant Human Protein MEMO1 protein (1-297AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together