Anti-CLCNKB Rabbit Recombinant polyclonal Antibody
77853-197
New Product
- Antigen Symbol:CLCNKB
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P51801
- Antigen Synonyms:Bartter syndrome type 3|Chloride channel Kb|Chloride channel kidney B|Chloride channel protein ClC-Kb|Chloride channel voltage sensitive Kb|ClC K2|ClC-K2|ClCK2|CLCKB|CLCKB_HUMAN|hClC Kb|hClCKb|MGC24087|OTTHUMP00000011120|OTTHUMP00000011121|RP11 5P18.8
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Chloride channel protein ClC-Kb protein (538-687AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:200-1:500, IF:1:50-1:200
Type:
Antigen: CLCNKB
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat